Order Entry
Spain
Orders LinkContactUsLinkComponent
Pitstop® 1 - negative control ≥95% Negative control for clathrin inhibitor Pitstop®1

Pitstop® 1 - negative control ≥95% Negative control for clathrin inhibitor Pitstop®1

Catalog # ABCAAB120686-5
Supplier:   Abcam
Pitstop® 1 - negative control ≥95% Negative control for clathrin inhibitor Pitstop®1
Catalog # ABCAAB120686-5
Supplier:  Abcam
CAS Number:  
Pitstop® 1 - negative control ≥95% Negative control for clathrin inhibitor Pitstop®1
CAS Number:  
Price
221,00 €
Log in for availability
1 * 5 mg
Product Details & Documents

Negative control for clathrin inhibitor Pitstop® 1 which is of the same chemical class and has a highly related structure to Pitstop® 1. Inhibits amphiphysin binding to the clathrin TD with an IC₅₀ 100 μm and does not block receptor-mediated endocytosis at concentrations of up to 300 μm (unpublished reports).

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Synonyms:  Pitstop 1™ - negative control
  • Molecular Weight:420,48 g/mol
Specifications

Specification Test Results

  • Constituents::
    100% Pitstop 1™ - negative control
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.