Order Entry
Spain
Orders LinkContactUsLinkComponent
 

Dodec-2-enoic acid ≥95% Diffusible fatty acid signal molecule

Catalog # ABCAAB145341-10
Supplier:   Abcam
Dodec-2-enoic acid ≥95% Diffusible fatty acid signal molecule
Catalog # ABCAAB145341-10
Supplier:  Abcam
CAS Number:  4412-16-2
Dodec-2-enoic acid ≥95% Diffusible fatty acid signal molecule
CAS Number:  4412-16-2
Price
298,00 €
Log in for availability
1 * 10 mg
Product Details & Documents

Diffusible fatty acid signal molecule. Acts as a bacterial virulence factor. Activates DSF-dependent responses. Inhibits germ tube formation in Candida albicans. Shows antifungal effects in vivo..

 : All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

  • CAS Number:4412-16-2
  • Molecular Weight:198,3 g/mol
Specifications

Specification Test Results

  • Constituents::
    100% cis-2-Dodecenoic acid