Order Entry
Spain
Orders LinkContactUsLinkComponent
 

DAF-2 diacetate ≥98% fluorescent probe, for nitric oxide detection

Supplier:   Abcam
DAF-2 diacetate ≥98% fluorescent probe, for nitric oxide detection
Supplier:  Abcam
Certificates
DAF-2 diacetate ≥98% fluorescent probe, for nitric oxide detection

2 Options Available Below

Product Details & Documents

Cell-permeable fluorescent nitric oxide (NO) indicator. Deacetylated by intracellular esterases to DAF-2. DAF-2 reacts with NO to yield the highly fluorescent triazolofluorescein (DAF-2T).

 : All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Synonyms:  2-​(3,​6-​diacetyloxy-​4,​5-​diamino-​9H-​xanthen-​9-​yl)-​benzoic acid, DAF-2 DA, 4,5-Diaminofluorescein diacetate
  • CAS Number:205391-02-2
  • Molecular Weight:446,41 g/mol
Specifications for Product Family

Specification Test Results

  • Constituents::
    100% DAF-2 DA
Product Family Options

Product Information

  • Availability
    Price
  • Log in for availability
    Log in for availability
    1 * 1 mg
    1 * 1 mg - 446,00 €
    446,00 €
    1 * 1 mg
       446,00 €

  • Log in for availability
    Log in for availability
    1 * 5 mg
    1 * 5 mg - 1 750,00 €
    1 750,00 €
    1 * 5 mg
       1 750,00 €