NANOCOLOR® Photometric Tube Test Kit for Starch
MANA985085
:
- Chemical method:Iodine-Starch reaction
- Pk:1 KIT
- Description:NANOCOLOR® Starch 100, test no. 0-85
- Dimensions:160×110×119 mm (including packaging)
- Model:MANA985085
- Platform:NANOCOLOR® tube tests
- Storage temperature:15…25 °C
- Shelf life (months):12
- Weight:383 g (including packaging)
- For unit:UV/VIS II; VIS II; 500 D; PF-12Plus; Advance
- Tests per kit:19
- Range:5 - 100 mg/L (Starch)
Photometric test kit for the determination of starch in water samples.
- Measurement directly in the test tube, saving time and reducing exposure to chemical reagents
- Bar code cuvette identification for easy test call up in NANOCOLOR® spectrophotometers
- Precisely pre-dosed reagents with constant high quality from batch to batch
- Coloured pictogram instructions and easy handling of the tubes
- 16 mm (OD) tubes for easy pipetting
The test kit is evaluated in 16 mm (OD) tubes. It is preprogrammend on NANOCOLOR® photometers. Programming data for other spectrophotometers are available.
Sea water analysis: Yes
Hazardous material: Yes
Remark: Measuring range on NANOCOLOR VIS II
: Rugged box with 19 test tubes.