Phospho-Src (Y419) Quantitative ELISA Kit
Catalog # ABCAAB279937-1X96
Supplier: Abcam
CAS Number:
Specifications
- Assay duration:Multiple steps
- Format:Pre-coated
- Target protein:Src
- Description:Phospho-Src (Y419) Quantitative ELISA kit
- Sample type:Cell lysate
- Storage temperature:4 °C
- Regulatory status:RUO
- Tests per kit:96 tests
Specifications
About this item
Phospho-Src (Y419) Quantitative ELISA kit is a Quantitative ELISA kit for the measurement of Phospho-Src (Y419) Quantitative in Human in Cell Lysate samples.