Mouse S100A8 ELISA Kit
Catalog # ABCAAB213886-96
Supplier: Abcam
CAS Number:
Specifications
- Description:Mouse S100A8 ELISA kit
Specifications
About this item
Mouse S100A8 ELISA kit is a Sandwich (quantitative) ELISA kit for the measurement of Mouse S100A8 in Mouse in cell Lysate, cell culture supernatant, EDTA plasma, Heparin plasma, serum samples.