Order Entry
Spain
Orders LinkContactUsLinkComponent
Human Complement Factor B (Ba fragment) ELISA Kit

Human Complement Factor B (Ba fragment) ELISA Kit

Human Complement Factor B (Ba fragment) ELISA kit, 96 tests
Catalog # ABCAAB315062-96TST
New Product
Supplier:   Abcam
Human Complement Factor B (Ba fragment) ELISA Kit
Catalog # ABCAAB315062-96TST
Supplier:  Abcam
New Product
CAS Number:  
Human Complement Factor B (Ba fragment) ELISA Kit
Human Complement Factor B (Ba fragment) ELISA Kit

Human Complement Factor B (Ba fragment) ELISA kit, 96 tests

CAS Number:  
Price
790,00 €
Log in for availability
1 * 1 Tests
Product Details & Documents

Human Complement Factor B (Ba fragment) ELISA Kit is a single-wash 90-min Simplestep used to quantify Human Complement Factor B (Ba fragment) with a sensitivity of 12,9 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader.

Specifications

Specifications

  • Description:
    Human Complement Factor B (Ba fragment) ELISA kit
  • Tests per kit:
    96 tests
  • Primary antibody reactivity:
    Human
  • Sample type:
    Serum, EDTA Plasma, Heparin Plasma, Cell culture supernatant
  • Detection range:
    21,875 - 1400 pg/ml
  • Storage temperature:
    +4 °C
  • Assay Type:
    Sandwich
  • Time to Results:
    1 h 30 min
  • Cat. No.:
    ABCAAB315062-96TST
  • Assay duration:
    One step
  • Detection method:
    Colorimetric
  • Sensitivity:
    12,9 pg/ml
  • Format:
    Pre-coated
  • Target protein:
    Complement Factor B
  • Regulatory status:
    RUO

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.