Order Entry
Spain
Orders LinkContactUsLinkComponent
Human CD62E ELISA Set (without plates)

Human CD62E ELISA Set (without plates)

Supplier:   Abcam
Human CD62E ELISA Set (without plates)
Supplier:  Abcam
Certificates
Human CD62E ELISA Set (without plates)

2 Options Available Below

Product Details & Documents

Human CD62E ELISA Set (without plates) is a Sandwich (quantitative) ELISA kit for the measurement of Human CD62E ELISA Set (without plates) in Human in Plasma, Cell culture supernatant, Serum samples.

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • ABCAAB47348-480
  • ABCAAB47348-10X96
  • Target protein:
  • CD62E
  • CD62E
  • Primary antibody reactivity:
  • Human
  • Human
  • Assay Type:
  • Sandwich
  • Sandwich
  • Format:
  • Pre-coated
  • Pre-coated
  • Assay duration:
  • Multiple steps
  • Multiple steps
  • Sensitivity:
  • 0,5 ng/ml
  • 0,5 ng/ml
  • Detection range:
  • 1 - 32 ng/ml
  • 1 - 32 ng/ml
  • Detection method:
  • Colorimetric
  • Colorimetric
  • Sample type:
  • Plasma, Cell culture supernatant, Serum
  • Plasma, Cell culture supernatant, Serum
  • Time to Results:
  • 2 h 15 min
  • 2 h 15 min
  • Regulatory status:
  • RUO
  • RUO
  • Storage temperature:
  • 4 °C
  • 4 °C

Specifications

Product Family Options

Product Information

  • Tests per kit
    Availability
    Price
  • ABCAAB47348-480
    Human CD62E ELISA Set (without plates)
    Human CD62E ELISA Set (without plates)
    5×96 tests
    Log in for availability
    Log in for availability
    1 * 480 Tests
    1 * 480 Tests - 938,00 €
    938,00 €
    1 * 480 Tests
       938,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    ABCAAB47348-480
    Target protein
    CD62E
    Primary antibody reactivity
    Human
    Assay Type
    Sandwich
    Format
    Pre-coated
    Assay duration
    Multiple steps
    Sensitivity
    0,5 ng/ml
    Detection range
    1 - 32 ng/ml
    Detection method
    Colorimetric
    Sample type
    Plasma, Cell culture supernatant, Serum
    Time to Results
    2 h 15 min
    Regulatory status
    RUO
    Storage temperature
    4 °C
  • ABCAAB47348-10X96
    Human CD62E ELISA Set (without plates)
    Human CD62E ELISA Set (without plates)
    10×96 tests
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT - 1 140,00 €
    1 140,00 €
    1 * 1 KIT
       1 140,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    ABCAAB47348-10X96
    Target protein
    CD62E
    Primary antibody reactivity
    Human
    Assay Type
    Sandwich
    Format
    Pre-coated
    Assay duration
    Multiple steps
    Sensitivity
    0,5 ng/ml
    Detection range
    1 - 32 ng/ml
    Detection method
    Colorimetric
    Sample type
    Plasma, Cell culture supernatant, Serum
    Time to Results
    2 h 15 min
    Regulatory status
    RUO
    Storage temperature
    4 °C