Mouse Galectin 3 ELISA Kit
Catalog # ABCAAB113343-96
Supplier: Abcam
CAS Number:
Specifications
- Description:Mouse Galectin 3 ELISA kit
Specifications
About this item
Mouse Galectin 3 ELISA kit is a Sandwich (quantitative) ELISA kit for the measurement of Mouse Galectin 3 in Mouse in plasma, cell culture supernatant, serum samples.