Order Entry
Spain
ContactUsLinkComponent
Diluent M Concentrate (10x)
Diluent M Concentrate (10x)
Catalog # ABCAAB138908-30
Supplier:  Abcam
CAS Number:  
undefined
Diluent M Concentrate (10x)
Catalog # ABCAAB138908-30
Supplier:  Abcam
CAS Number:  

Specifications

  • Description:
    Diluent M concentrate (10X)
  • Size:
    30 ml
  • Storage temperature:
    4 °C
  • Plate types:
    Pre-coated

Specifications

About this item

Diluent M Concentrate (10X) (ab138908) is part of the reagents, controls & accessories range. Abcam offers high-quality biological reagents and tools including antibodies, proteins, assays, cell lines and lysates.

Caution:

Research use only.