Order Entry
Spain
ContactUsLinkComponent
Rabbit MMP9 ELISA Kit
Rabbit MMP9 ELISA Kit
Catalog # ABCAAB273243-1X96
Supplier:  Abcam
CAS Number:  
undefined
Rabbit MMP9 ELISA Kit
Catalog # ABCAAB273243-1X96
Supplier:  Abcam
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type:
    Sandwich
  • Format:
    Pre-coated
  • Target protein:
    MMP9
  • Description:
    Rabbit MMP9 ELISA kit
  • Sample type:
    Plasma, cell culture supernatant, serum
  • Detection method:
    Colorimetric
  • Detection range:
    61,44 - 15000 pg/ml
  • Storage temperature:
    4 °C
  • Sensitivity:
    61 pg/ml
  • Regulatory status:
    RUO
  • Tests per kit:
    96 tests

Specifications

About this item

Rabbit MMP9 ELISA kit is a Sandwich (quantitative) ELISA kit for the measurement of Rabbit MMP9 in Rabbit in plasma, cell culture supernatant, Serum samples.