Mouse IgG2c ELISA Kit
Mouse IgG2c ELISA Kit, 96 tests
ABCAAB157720-96
: Abcam
Mouse IgG2c ELISA kit is a Sandwich (quantitative) ELISA kit for the measurement of Mouse IgG2c in Mouse in plasma, serum samples.
- Description:Mouse IgG2c ELISA Kit
- Tests per kit:96 tests
- Cat. No.:ABCAAB157720-96
- Assay duration:Multiple steps
- Detection method:Colorimetric
- Sensitivity:0,38 ng/ml
- Format:Pre-coated
- Primary antibody reactivity:Mouse
- Sample type:Plasma, Serum
- Target protein:IgG2c
- Detection range:3,13 - 200 ng/ml
- Storage temperature:4 °C
- Regulatory status:RUO
- Assay Type:Sandwich



