Phosphotyrosine Alpha-synuclein Quantitative ELISA Kit
Catalog # ABCAAB279935-1X96
Supplier: Abcam
CAS Number:
Specifications
- Descripción:Phosphotyrosine Alpha-synuclein Quantitative ELISA kit
- Temperatura de almacenamiento:+4 ℃
- Tests por kit:96 tests
Specifications
About this item
Phosphotyrosine Alpha-synuclein Quantitative ELISA kit is a Quantitative ELISA kit for the measurement of Phosphotyrosine Alpha-synuclein Quantitative in Human in cell lysate samples.