Mouse ARP1 ELISA Kit
Catalog # ABCAAB267590-1X96
Supplier: Abcam
CAS Number:
Specifications
- Description:Mouse ARP1 ELISA kit
- Storage temperature:+4 ℃
- Tests per kit:96 tests
Specifications
About this item
Mouse ARP1 ELISA kit is a Sandwich (quantitative) ELISA kit for the measurement of Mouse ARP1 in Mouse in plasma, cell culture supernatant, serum samples.