Human ANGPTL7 ELISA Kit
ABCAAB313507-96TST
: Abcam
:
- Description:Human ANGPTL7 ELISA Kit
- Tests per kit:96 tests
Human ANGPTL7 ELISA Kit is a Sandwich (quantitative) ELISA kit for the measurement of Human ANGPTL7 in Human in Cell culture supernatant, Plasma, Serum samples.