Mouse Endocan ELISA Kit
Catalog # ABCAAB193722-96
Supplier: Abcam
CAS Number:
Specifications
- Description:Mouse Endocan ELISA Kit
- Tests per kit:96 tests
Specifications
About this item
Mouse Endocan ELISA Kit is a Sandwich (quantitative) ELISA kit for the measurement of Mouse Endocan in Mouse in Plasma, Cell culture media, Serum samples.