Phospho-JAK1 (Y1022) ELISA Kit
Catalog # ABCAAB279837-1X96
Supplier: Abcam
CAS Number:
Specifications
- Description:Phospho-JAK1 (Y1022) ELISA kit
- Tests per kit:96 tests
Specifications
About this item
Phospho-JAK1 (Y1022) ELISA kit is a Semi-quantitative ELISA kit for the measurement of Phospho-JAK1 (Y1022) in Human in cell lysate samples.