Mouse Albumin ELISA Kit
Catalog # ABCAAB108792-96
Supplier: Abcam
CAS Number:
Specifications
- Description:Mouse Albumin ELISA kit
- Tests per kit:96 tests
Specifications
About this item
Mouse Albumin ELISA Kit is a Sandwich ELISA used to quantify Mouse Albumin with a sensitivity of 0,48 ng/ml - Colorimetric ELISA kit - 450 nm readout. Works on any standard plate reader - Wide dynamic range – 0,781 - 200 ng/ml - Cited in over 95 citations.