Human HSD17B1 ELISA Kit
Catalog # ABCAAB283975-1X96
Supplier: Abcam
CAS Number:
Specifications
- Description:Human HSD17B1 ELISA Kit
- Tests per kit:96 tests
Specifications
About this item
Human HSD17B1 ELISA Kit is a Sandwich (quantitative) ELISA kit for the measurement of Human HSD17B1 in Human in Plasma, Cell culture supernatant, Serum samples.