Human TPFI2 ELISA Kit
Catalog # ABCAAB275908-1X96
Supplier: Abcam
CAS Number:
Specifications
- Description:Human TPFI2 ELISA kit
Specifications
About this item
Human TPFI2 ELISA kit is a single-wash 90-min Simplestep used to quantify Human TPFI2 with a sensitivity of 16 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. Colorimetric Sandwich ELISA - 450 nm readout: works on any standard plate reader.