Rabbit IL-4 ELISA Kit
Catalog # ABCAAB273244-1X96
Supplier: Abcam
CAS Number:
Specifications
- Description:Rabbit IL-4 ELISA kit
- Tests per kit:96 tests
Specifications
About this item
Rabbit IL-4 ELISA kit is a sandwich (quantitative) ELISA kit for the measurement of Rabbit IL-4 in Rabbit in Plasma, cell culture supernatant, serum samples.