Mouse IFNGR1 ELISA Kit
Catalog # ABCAAB267580-1X96
Supplier: Abcam
CAS Number:
Specifications
- Description:Mouse IFNGR1 ELISA kit
- Tests per kit:96 tests
Specifications
About this item
Mouse IFNGR1 ELISA Kit is a Sandwich (quantitative) ELISA kit for the measurement of Mouse IFNGR1 in Mouse in Plasma, Cell culture media, Serum samples.