Cow IgM ELISA Kit
Catalog # ABCAAB190518-96
Supplier: Abcam
CAS Number:
Specifications
- Description:Cow IgM ELISA kit
- Tests per kit:96 tests
Specifications
About this item
Cow IgM ELISA kit is a sandwich ELISA kit for the measurement of Cow IgM in Cow in Plasma, serum samples.
Certifications: RUO