Dimethyl-2,6-pyridinedicarboxylate ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:5453-67-8
:
Formula:C₉H₉NO₄
MDL Number:MFCD00134493
:195.17 g/mol
EINECS:226-697-3
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:121.0 to 126.0 deg-C
Specification Test Results
Frequently Bought Together