


1-Butanol ≥99.0% (by GC) for spectrophotometry
TCIAB0228-100ML
: TCI
:
- Pack type:Glass bottle
- Pk:100 mL
- Seq:2440
- Appearance:Colorless clear liquid
- Purity (GC):min. 99.0 %
- Density (20deg.C):0.8080 to 0.8120 g/mL
- Absorbance (210nm):max. 1.000
- Absorbance (220nm):max. 0.500
:
:74.12 g/mol
:14,01540
:118°C
:–89 °C
:0,812 g/cm³ (20 °C)
:37°C
:Ambient
:200-751-6
:1120
:3,III
:01-2119484630-38
:GHS02
:GHS05
:GHS07
:71-36-3
:H₃C(CH₂)₂CH₂OH
:MFCD00002964