Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Spain
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Sodium dodecyl sulphate (SDS)
Sodium dodecyl sulphate (SDS) 200 g/l in aqueous solution
Sodium dodecyl sulphate (SDS) 200 g/l in aqueous solution
PRLS88711.320
:
PROLABO CMP
:
Sodium dodecyl sulphate (SDS) 200 g/l in aqueous solution
PRLS88711.320
:
PROLABO CMP
:
Pack type:
Glass bottle
Pk:
2,5 L
Seq:
5096
:
:
288.38 g/mol
:
1…1.1 g/cm³ (20 °C)
:
Ambient
:
205-788-1
:
151-21-3
:
CH₃(CH₂)₁₁OSO₃Na
:
MFCD00036175