
3-Methylcyclohexanol cis- and trans-mixture ≥98.0% (by GC)
: TCI
:591-23-1
:
:C₇H₁₄O
:MFCD00001446
:114.19 g/mol
:171 - 173 °C (1013 hPa)
:–74 °C
:0,9158 g/cm³ (20 °C)
:62°C
:209-709-1
:GHS07
:Ambient
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 % (total of isomers)
Frequently Bought Together