Order Entry
Spain
ContactUsLinkComponent
Fast green FCF ≥85.0% (by HPLC and UV method) for biochemical research
Fast green FCF ≥85.0% (by HPLC and UV method) for biochemical research
Catalog # TCIAF0718-5G
Supplier:  TCI
CAS Number:  
undefined
Fast green FCF ≥85.0% (by HPLC and UV method) for biochemical research
Catalog # TCIAF0718-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    Orange to Brown to Dark purple powder to crystal
  • Purity (HPLC):
    min. 85.0 area%
  • Purity (UV-vis method):
    min. 85.0 % (calcd.on anh.substance)
  • Lambda max.:
    622.0 to 626.0 nm (0.02mol/L AcONH4 sol.)
  • Drying loss:
    max. 10.0 %

Specification Test Results

About this item

 :
 :808.86 g/mol
Merck Index:14,03941
Melting Pt:290 °C
EINECS:219-091-5
 :GHS08
Storage Temperature:Ambient
CAS Number:2353-45-9
Formula:C₃₇H₃₄N₂Na₂O₁₀S₃
MDL Number:MFCD00013053