Order Entry
Spain
ContactUsLinkComponent
 
 
 
 
N,N-Dimethylformamide ≥99.5% (by GC) for spectrophotometry
 :  TCI
 
undefined
 
 
 
N,N-Dimethylformamide ≥99.5% (by GC) for spectrophotometry
 :  TCI

 

  • Appearance:
    Colorless clear liquid
  • Purity (GC):
    min. 99.5 %
  • Density (20deg.C):
    0.9470 to 0.9520 g/mL
  • Absorbance (270nm):
    max. 0.80
  • Absorbance (280nm):
    max. 0.40

 

Frequently Bought Together