4-Methyl-2-pentene cis- and trans-mixture ≥96.0% (by GC)
: TCI
:4461-48-7
:
:C₆H₁₂
:MFCD00065138
:84.16 g/mol
:58 °C (1013 hPa)
:–134.4 °C
:0,6664 g/cm³ (20 °C)
:–32°C
:224-721-7
:2288
:3,II
:GHS02
:GHS08
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 96.0 % (total of isomer)
Frequently Bought Together