Order Entry
Spain
ContactUsLinkComponent
Crotyl alcohol cis- and trans-mixture ≥95.0% (by GC)
Crotyl alcohol cis- and trans-mixture ≥95.0% (by GC)
Catalog # TCIAB0696-25ML
Supplier:  TCI
CAS Number:  
undefined
Crotyl alcohol cis- and trans-mixture ≥95.0% (by GC)
Catalog # TCIAB0696-25ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 mL
  • Seq:
    2315
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 95.0 % (total of isomer)

Specification Test Results

About this item

Synonyms: 2-Buten-1-ol, 1-Hydroxy-2-butene
 :
 :72.11 g/mol
EINECS:228-086-7
UN:1987
 :GHS02
 :GHS07
Storage Temperature:Ambient
CAS Number:6117-91-5
Formula:C₄H₈O
MDL Number:MFCD00002923
Boiling Pt:121°C
Flash Pt:34°C
ADR:3 III
Merck Index:14,02601