
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) ≥99.0% (by titrimetric analysis) for electrophoresis
Supplier: TCI
Certificates
About this item
CAS Number:77-86-1
:
Formula:NH₂C(CH₂OH)₃
MDL Number:MFCD00004679
:121.14 g/mol
Merck Index:14,09772
Boiling Pt:220°C
Melting Pt:169…172 °C
Density:1,328 g/cm³ (20 °C)
EINECS:201-064-4
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Neutralization titration):min. 99.0 %
- Functionality test:to pass test
Specification Test Results
Frequently Bought Together