Iodoacetamide ≥98.0% (by total nitrogen basis) for biochemical research
Catalog # TCIAI0741-5G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (with Total Nitrogen):min. 98.0 %
- Melting point:93.0 and le 97.0 deg-C
- Enzyme inhibition test:to pass test
Specification Test Results
About this item
Synonyms: 2-Iodoacetamide
:
:184.96 g/mol
Boiling Pt:266 °C (1013 hPa)
Melting Pt:91…94 °C
Storage Temperature:Refrigerator
EINECS:205-630-1
UN:2811
ADR:6.1,III
:GHS06
CAS Number:144-48-9
Formula:ICH₂CONH₂
MDL Number:MFCD00008028