Entrada de pedido
Spain
Orders LinkContactUsLinkComponent
General-purpose thermometer
General-purpose thermometer
−10…+250 °C, NS 14/23, Blue spirit
Catalog # 620-2290
Supplier:   SciLabware
General-purpose thermometer
Catalog # 620-2290
Supplier:  SciLabware
General-purpose thermometer
−10…+250 °C, NS 14/23, Blue spirit
Price
192,00 €
Log in for availability
1 * 1 UN
Product Details & Documents
Spirit thermometers.
An integral ground glass cone on the stem allows direct fitting into Quickfit® jointed apparatus. Spirit thermometers have blue or green spirit for improved visibility.
Specifications
  • Seq:
    01
  • División:
    NS 14/23
  • Relleno:
    Blue spirit
  • Rango:
    −10…+250 °C