Order Entry
Spain
ContactUsLinkComponent
Sulphur trioxide pyridine complex ≥95.0% (by titrimetric analysis)
Corrosive
Irritant
Sulphur trioxide pyridine complex ≥95.0% (by titrimetric analysis)
Supplier:  TCI
Certificates
undefined
Corrosive
Irritant
Sulphur trioxide pyridine complex ≥95.0% (by titrimetric analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Light yellow powder to crystal
  • Purity (Ethanolysis Method):
    min. 95.0 %

Specification Test Results

Related Information

Frequently Bought Together