Order Entry
Spain
ContactUsLinkComponent
 
 
 
 
2,4-Dichlorophenol ≥98.0% (by GC) for biochemical research
 :  TCI
 
undefined
 
 
 
2,4-Dichlorophenol ≥98.0% (by GC) for biochemical research
 :  TCI

 

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Melting point:
    40.0 to 44.0 deg-C
  • Hydrogendioxide detection test (Peroxidase method):
    to pass test

 

Frequently Bought Together