



n-Octane ≥98.5% (by GC) for HPLC
Catalog # TCIAO0151-500ML
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:500 mL
- Seq:2725
Specifications
Specification Test Results
- Appearance:Colorless clear liquid
- Purity (GC):min. 98.5 %
- Absorbance (210nm):max. 0.350
- Absorbance (220nm):max. 0.150
- Absorbance (230nm):max. 0.060
Specification Test Results
About this item
:
:114.23 g/mol
Merck Index:14,06749
Boiling Pt:125°C
Melting Pt:-57°C
Melting Pt:–57°C
Density:0,703 g/cm³ (20 °C)
Flash Pt:14°C
Storage Temperature:Ambient
EINECS:203-892-1
UN:1262
ADR:3,II
:GHS02
:GHS07
:GHS08
:GHS09
CAS Number:111-65-9
Formula:H₃C(CH₂)₆CH₃
MDL Number:MFCD00009556