Order Entry
Spain
ContactUsLinkComponent
 
Flammable
Irritant
Health
Environment
n-Hexane, anhydrous ≥96.0% (by GC)
Catalog # TCIAH1197-500ML
Supplier:  TCI
CAS Number:  
undefined
Flammable
Irritant
Health
Environment
n-Hexane, anhydrous ≥96.0% (by GC)
Catalog # TCIAH1197-500ML
Supplier:  TCI
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Seq:
    2725

Specifications

Specification Test Results

  • Appearance:
    Colorless clear liquid
  • Purity (GC):
    min. 96.0 %
  • Water:
    max. 50 ppm

Specification Test Results

About this item

 :
 :86.18 g/mol
Merck Index:14,04694
Boiling Pt:68°C
Melting Pt:–95°C
Density:0,659 g/cm³ (20 °C)
Flash Pt:–22°C
Storage Temperature:Ambient
EINECS:203-777-6
UN:1208
ADR:3,II
 :GHS02
 :GHS07
 :GHS08
 :GHS09
CAS Number:110-54-3
 :92112-69-1
Formula:H₃C(CH₂)₄CH₃
MDL Number:MFCD00009520