Order Entry
Spain
ContactUsLinkComponent
Silver nitrate 10% in aqueous solution
Corrosive
Environment
Silver nitrate 10% in aqueous solution
Catalog # PRLSMC306815.1000
Supplier:  PROLABO CMP
CAS Number:  
undefined
Corrosive
Environment
Silver nitrate 10% in aqueous solution
Catalog # PRLSMC306815.1000
Supplier:  PROLABO CMP
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :169.87 g/mol
EINECS:231-853-9
 :GHS05
 :GHS09
Storage Temperature:Ambient
CAS Number:7761-88-8
Formula:AgNO₃
MDL Number:MFCD00003414