2-Thiobarbituric acid ≥98.0% (by titrimetric analysis) for biochemical research
Catalog # TCIAT2834-5G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (Neutralization titration):min. 98.0 %
- TBARS test:to pass test
Specification Test Results
About this item
:
:144.15 g/mol
Boiling Pt:419 °C (1013 hPa)
Melting Pt:245°C
Density:1,661 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:207-985-8
CAS Number:504-17-6
Formula:C₄H₄N₂O₂S
MDL Number:MFCD00006674