Order Entry
Spain
ContactUsLinkComponent
Methyl-1-naphthoate ≥98.0% (by GC)
Methyl-1-naphthoate ≥98.0% (by GC)
Catalog # TCIAN0615-25G
Supplier:  TCI
CAS Number:  
undefined
Methyl-1-naphthoate ≥98.0% (by GC)
Catalog # TCIAN0615-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    Colorless to Yellow to Green clear liquid
  • Purity (GC):
    min. 98.0 %

Specification Test Results

About this item

 :
 :186.2066 g/mol
Boiling Pt:169°C
CAS Number:2459-24-7
Formula:C₁₂H₁₀O₂
MDL Number:MFCD00094243