
3,5-Dichlorosulphanilamide ≥98.0% (by total nitrogen basis)
Supplier: TCI
Certificates
About this item
CAS Number:22134-75-4
:
Formula:C₆H₆Cl₂N₂O₂S
MDL Number:MFCD00014784
:241.10 g/mol
EINECS:244-799-6
:GHS07
Specification Test Results
- Appearance:Light yellow to Brown powder to crystal
- Purity (GC):min. 96.0 %
- Purity (with Total Nitrogen):min. 98.0 %
- Melting point:205.0 to 208.0 deg-C
- Solubility in hot Methanol:almost transparency
Specification Test Results
Frequently Bought Together