
(4-Aminophenyl)acetonitrile ≥98.0% (by GC, titration analysis)
: TCI
:3544-25-0
:
:C₈H₈N₂
:MFCD00007912
:132.17 g/mol
:Refrigerator
:222-587-4
:3439
:6.1,III
:GHS06
- Appearance:White or Colorless to Brown powder to lump to clear liquid
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:42.0 to 46.0 deg-C
Frequently Bought Together