Order Entry
Spain
ContactUsLinkComponent
Alizarine yellow R ≥98.0% (by HPLC)
Alizarine yellow R ≥98.0% (by HPLC)
Catalog # TCIAA0579-25G
Supplier:  TCI
CAS Number:  
undefined
Alizarine yellow R ≥98.0% (by HPLC)
Catalog # TCIAA0579-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    Light yellow to Amber to Dark green powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Absorbance:
    min. 0.320 (5 mg/L, pH 9.6, 375.0 nm) (calcd.on dried substance)
  • Drying loss:
    max. 10.0 %
  • Transition interval (pH):
    (redish yellow)10.0 to 12.0 (yellowish red)

Specification Test Results

About this item

 :
Merck Index:14,00255
EINECS:218-818-3
CAS Number:2243-76-7
Formula:C₁₃H₉N₃O₅
MDL Number:MFCD00007313