Triethyl-1,1,2-ethanetricarboxylate ≥96.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:7459-46-3
:
Formula:C₁₁H₁₈O₆
MDL Number:MFCD00009154
:246.26 g/mol
Boiling Pt:165°C
EINECS:231-235-9
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 96.0 %
Specification Test Results
Frequently Bought Together