Order Entry
Spain
ContactUsLinkComponent
1,2,3,4-Tetrahydronaphthalen-2-one (2-tetralone-β-tetralone) ≥95.0% (by GC)
1,2,3,4-Tetrahydronaphthalen-2-one (2-tetralone-β-tetralone) ≥95.0% (by GC)
Supplier:  TCI
Certificates
undefined
1,2,3,4-Tetrahydronaphthalen-2-one (2-tetralone-β-tetralone) ≥95.0% (by GC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    Colorless to Light orange to Yellow clear liquid
  • Purity (GC):
    min. 95.0 %

Specification Test Results

Frequently Bought Together