Order Entry
Spain
ContactUsLinkComponent
1,1-Dimethoxycyclopentane ≥96.0% (by GC)
Flammable
1,1-Dimethoxycyclopentane ≥96.0% (by GC)
Supplier:  TCI
Certificates
undefined
Flammable
1,1-Dimethoxycyclopentane ≥96.0% (by GC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 96.0 %

Specification Test Results

Frequently Bought Together