Order Entry
Spain
ContactUsLinkComponent
 
(2-Bromophenyl)acetone ≥98.0% (by GC)
 :  TCI
 
undefined
(2-Bromophenyl)acetone ≥98.0% (by GC)
 :  TCI

 

  • Appearance:
    Light orange to Yellow to Green clear liquid
  • Purity (GC):
    min. 98.0 %

 

Frequently Bought Together