D-Panthenol ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:81-13-0
:
Formula:C₉H₁₉NO₄
MDL Number:MFCD00065006
:205.25 g/mol
Merck Index:14,02947
EINECS:201-327-3
:GHS07
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (Neutralization back titration):min. 98.0 %
- Specific rotation [a]20/D:+29.0 to +32.0 deg (C=5, H2O)
- Solubility in Water:almost transparency
Specification Test Results
Frequently Bought Together