(1S)-(+)-Camphorquinone ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:2767-84-2
:
Formula:C₁₀H₁₄O₂
MDL Number:MFCD00064159
:166.22 g/mol
EINECS:220-446-1
Specification Test Results
- Appearance:White to Yellow to Orange powder to crystal
- Purity (GC):min. 97.0 %
- Melting point:198.0 to 202.0 deg-C
- Specific rotation [a]20/D:+95.0 to +103.0 deg (C=1,Toluene)
Specification Test Results
Frequently Bought Together